Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum oxide, Alpha-phase, 99.9% (metals basis)

Catalog No. AA42572A3
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42572A3 2 kg
AA4257222 100 g
AA4257236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA42572A3 Supplier Thermo Scientific Chemicals Supplier No. 042572A3
Only null left
Add to Cart

S.A. 6-8m2/g | CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

Aluminum oxide is widely used in a variety of plastics, rubber, ceramics, refractory products for reinforcement toughening. It is also useful to produce artificial ruby, sapphire, yttrium aluminum garnet. It is useful in the field of fiber fabric products and high pressure sodium lamp as far-infrared emission and thermal insulation materials. It is useful for the C single crystal detectors. It plays an important role for the hydrogenation of aromatic hydrocarbons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Boiling Point 2980°C
Melting Point 2045°C
Color White
Physical Form Powder
Quantity 2 kg
Assay Percent Range 99.9% (Metal basis)
Solubility Information Slowly soluble in aqueous alkali solutions-forming hydroxides. Insoluble in water and in non-polar organic solvents.
IUPAC Name dialuminium(3+) trioxidandiide
Formula Weight 101.96
Odor Odorless
Refractive Index 1.765
Appearance White powder
Chemical Name or Material Aluminum oxide
Show More Show Less
EINECSNumber 215-691-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.