Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alumina, Calcined Aluminum Oxide, Certified, Fisher Chemical™

Catalog No. A591500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A591-500 500 g Glass Bottle
A591-3 3 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A591-500

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

  • Typical surface area 0.4 m2/g
  • Known for its insulative properties and hardness
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Boiling Point 2980°C
Melting Point 2000°C
Color White
Physical Form Solid
Quantity 500 g
Formula Weight 101.96
Grade Certified
Identification Pass Test
Loss on Ignition 1% max.
Packaging Glass Bottle
Chemical Name or Material Aluminum Oxide Anhydrous
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.