Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum oxide, NanoDur™ AL-2420, 50% in mineral spirits, colloidal dispersion with dispersant, Thermo Scientific™

Catalog No. AA4579022
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4579022 100 g
AA4579036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4579022 Supplier Thermo Scientific Chemicals Supplier No. 4579022

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Aluminum oxide dispersed in mineral spirits suitable for incorporation in non-polar organic based coatings and formulations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Physical Form Dry Powder
Quantity 100 g
Formula Weight 101.96
Concentration or Composition (by Analyte or Components) 50% in mineral spirits
Odor Mild
Chemical Name or Material Aluminum oxide, NanoDur™ AL-2420, Colloidal dispersion with dispersant
missing translation for 'healthHazard1' Danger
missing translation for 'healthHazard2' GHS H Statement
H304
May be fatal if swallowed and enters airways.
missing translation for 'healthHazard3' GHS P Statement
P301+P310-P331-P405-P501a
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.