Learn More
Aluminum oxide, NanoDur™ AL-2420, 50% in mineral spirits, colloidal dispersion with dispersant, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Aluminum oxide dispersed in mineral spirits suitable for incorporation in non-polar organic based coatings and formulations
Specifications
Specifications
| Physical Form | Dry Powder |
| Quantity | 500 g |
| Formula Weight | 101.96 |
| Concentration or Composition (by Analyte or Components) | 50% in mineral spirits |
| Odor | Mild |
| Chemical Name or Material | Aluminum oxide, NanoDur™ AL-2420, Colloidal dispersion with dispersant |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement H304 May be fatal if swallowed and enters airways. |
| Precautionary Statement | GHS P Statement P301+P310-P331-P405-P501a IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| TSCA | TSCA |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.