Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Oxide, Powder, BAKER ANALYZED™ Reagent, J.T.Baker™

Catalog No. 02002248 Shop All Avantor Products
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-002-248 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-002-248 Supplier Avantor Supplier No. 053605
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 1344-28-1
Physical Form Powder
Quantity 2.5 kg
Molecular Formula Al2O3
MDL Number MFCD00003424
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES [O--].[O--].[O--].[Al+3].[Al+3]
IUPAC Name dialuminium(3+) trioxidandiide
Molecular Weight (g/mol) 101.96
PubChem CID 9989226
Formula Weight 101.96
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Specific Gravity 1L = 4.00kg
Chemical Name or Material Aluminum Oxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.