Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum oxide, gamma-phase, 99.97% (metals basis)

Catalog No. p-3256863
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3981214 25 g
AA3981222 100 g
AA3981236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3981214 Supplier Thermo Scientific Chemicals Supplier No. 03981214
Only null left
Add to Cart

CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

Aluminum oxide, is useful to produce aluminum metal. It has many uses in pharmaceutical and industrial manufacturing processes as an adsorbent, desiccating agent, and catalyst, and in the manufacture of dental cements, available in toothpaste, as a dispersing agent food additive, and for uses in hemodialysis. Tailored aluminum oxide layers plays an important role for the bipolar current adjustment in the plasma electrolytic oxidation process. Red phosphorus/aluminum oxide composites play an essential role as flame retardants in recycled poly(ethylene terephthalate).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.97% (Metals basis)
Solubility Information Slightly soluble in cold water,acid and alkali. Insoluble in hot water.
Formula Weight 101.96
Odor Odorless
Chemical Name or Material Aluminum oxide, gamma
EINECSNumber 215-691-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.