Learn More
Cole-Parmer Cole-Parmer Aluminum Pan Liners, 1.2gr, 70mm I.D. (2 1/2" ) x 1/4" Depth, 500/Cs
Catalog No. NC1219830 Shop All Cole-Parmer Products
Encompass_Preferred
Weighing Pan, oil-free, ideal for general purpose weighing and dispensing. Compatible with both open and enclosed balances. Slightly tapered for stacking.
Catalog No. NC1219830 Supplier Cole-Parmer Supplier No. 0101820
Weighing Pan, oil-free, ideal for general purpose weighing and dispensing. Compatible with both open and enclosed balances. Slightly tapered for stacking.