Learn More
Thomas Scientific Aluminum PCR Plate Seal
SDP
Catalog No. NC1521799
Encompass
Adhesive PCR plate seals and cap strips prevent evaporation during thermal cycling or storage. Sealing mats can be reused up to five times and achieve complete sealing with screw/clip-down hot lid thermal cyclers. All PCR Plate and Tube sealers are certified RNase/DNase-Free, Human DNA/PCR Inhibitor-Free, and non-pyrogenic.
Catalog No. NC1521799 Supplier Thomas Scientific Supplier No. 1165R30
Adhesive PCR plate seals and cap strips prevent evaporation during thermal cycling or storage. Sealing mats can be reused up to five times and achieve complete sealing with screw/clip-down hot lid thermal cyclers. All PCR Plate and Tube sealers are certified RNase/DNase-Free, Human DNA/PCR Inhibitor-Free, and non-pyrogenic.