Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum perchlorate nonahydrate, 99%, extra pure, Thermo Scientific™

Catalog No. AC197171000
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC197171000 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC197171000 Supplier Thermo Scientific Chemicals Supplier No. 197171000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
CAS 81029-06-3
pH 0 to 3 (5% aq. soln.)
Physical Form Moist Crystals
Quantity 100 g
Assay Percent Range 99%
Molecular Formula AlCl3H18O21
Linear Formula Al(ClO4)3·9H2O
MDL Number MFCD00149130
Synonym aluminum perchlorate nonahydrate, al.3clo4.9h2o, aluminum perchlorate nonahydrate, reagent, aluminum 3+ nonahydrate triperchlorate
InChI Key CYRDFMGMNQJULY-UHFFFAOYSA-K
SMILES O.O.O.O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Al+3]
IUPAC Name aluminum;triperchlorate;nonahydrate
Molecular Weight (g/mol) 487.455
PubChem CID 16211522
Formula Weight 487.46
Percent Purity ≥98.5%
Grade Extra Pure
Chemical Name or Material Aluminum perchlorate nonahydrate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.