Learn More
Medchemexpress LLC Aluminum phthalocyanine chloride | 14154-42-8 | MFCD00049386 | 85.0% | 574.96 g/mol | C32H16AlClN8 | 5 MG
SDP
Catalog No. 5000262848
Encompass_Preferred
Aluminum phthalocyanine chloride is a photosensitizing phthalocyanine dye used in photodynamic therapy research and fluorescence studies. It is a chloroaluminum complex with strong absorption in the red region and is supplied as a research-grade solid with supporting documentation for laboratory use.
  • Photosensitizer with strong absorption in the red region.
  • Suitable for photodynamic therapy and fluorescence research applications.
  • Available in research-scale pack sizes for small-batch experiments.
  • Provided with safety data sheet and certificate of analysis for safe handling.
  • Stable solid form for storage and experimental reproducibility.
Catalog No. 50-002-62848 Supplier Medchemexpress LLC Supplier No. HY1299595MG
Aluminum phthalocyanine chloride is a photosensitizing phthalocyanine dye used in photodynamic therapy research and fluorescence studies. It is a chloroaluminum complex with strong absorption in the red region and is supplied as a research-grade solid with supporting documentation for laboratory use.
  • Photosensitizer with strong absorption in the red region.
  • Suitable for photodynamic therapy and fluorescence research applications.
  • Available in research-scale pack sizes for small-batch experiments.
  • Provided with safety data sheet and certificate of analysis for safe handling.
  • Stable solid form for storage and experimental reproducibility.