Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aluminum potassium sulfate | 10043-67-1 | MFCD00003428 | 99.6% | 258.21 g/mol | KAl(SO4)2 | 50 G
SDP

Catalog No. 5000282990
Encompass_Preferred

Aluminum potassium sulfate (potassium alum) is an inorganic salt used as a coagulant and antibacterial agent in laboratory research. It is supplied as a high-purity reagent for microbiology and environmental science applications.

  • High purity: 99.6%.
  • Chemical formula: KAl(SO4)2.
  • Molecular weight: 258.21 g/mol.
  • Applications: water purification and antibacterial research.
  • Pack size: 50 G laboratory reagent.
  • For research use only; not for clinical or human use.

Catalog No. 50-002-82990 Supplier Medchemexpress LLC Supplier No. HYW13430150G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aluminum potassium sulfate (potassium alum) is an inorganic salt used as a coagulant and antibacterial agent in laboratory research. It is supplied as a high-purity reagent for microbiology and environmental science applications.

  • High purity: 99.6%.
  • Chemical formula: KAl(SO4)2.
  • Molecular weight: 258.21 g/mol.
  • Applications: water purification and antibacterial research.
  • Pack size: 50 G laboratory reagent.
  • For research use only; not for clinical or human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.