Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Potassium Sulfate ACS AR Granular, Macron Fine Chemicals™

Catalog No. MK3216500 Shop All Avantor Products
Change view
Click to view available options
POLYSTORMOR; 12 x 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-3216-500 POLYSTORMOR; 12 x 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-3216-500 Supplier Avantor Supplier No. 321604

Specifications

CAS 7784-24-9
Physical Form Granules
Quantity 12 x 500 g
Molecular Formula AlH24KO20S2
Synonym kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrate
InChI Key GNHOJBNSNUXZQA-UHFFFAOYSA-J
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+]
IUPAC Name aluminum;potassium;disulfate;dodecahydrate
Molecular Weight (g/mol) 474.372
PubChem CID 62667
ChEBI CHEBI:86465
Formula Weight 474.38
Grade ACS
Packaging POLYSTORMOR
Chemical Name or Material Aluminum Potassium Sulfate, 12-Hydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.