Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Potassium Sulfate Dodecahydrate (Powder/USP/FCC), Fisher Chemical™

Catalog No. A605212 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A605-212 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A605-212

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Chemical Identifiers

CAS 7784-24-9
Molecular Formula AlH24KO20S2
Molecular Weight (g/mol) 474.372
MDL Number MFCD00149143
InChI Key GNHOJBNSNUXZQA-UHFFFAOYSA-J
Synonym kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrate
PubChem CID 62667
ChEBI CHEBI:86465
IUPAC Name aluminum;potassium;disulfate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+]

Specifications

Melting Point 92°C
Color White
pH 3.0 to 3.5
Physical Form Solid
Quantity 2.5 kg
Formula Weight 474.39
Percent Purity 99.0 to 100.5% (USP)
Grade FCC/USP
Identification Pass Test (FCC), Pass Test (USP), Pass Test (USP/FCC)
Loss on Drying 43.0 to 46.0% (USP)
Packaging Poly Bottle
Chemical Name or Material Aluminum Potassium Sulfate Dodecahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.