Learn More
Perkin Elmer US LLC Aluminum Probe Handle
Catalog No. 502112756
Encompass_Preferred
The Aluminum Probe Handle is used with screw-based probes. This handle comes standard with the Microsampling Tool Kit (N1870151). With this part number, this part can be ordered as a replacement or spare.
Catalog No. 50-211-2756 Supplier Perkin Elmer US LLC Supplier No. N9302622
The Aluminum Probe Handle is used with screw-based probes. This handle comes standard with the Microsampling Tool Kit (N1870151). With this part number, this part can be ordered as a replacement or spare.