Learn More
McMaster-Carr Multipurpose 6061 Aluminum Rectangular Tube
Catalog No. NC2899605
Encompass
Multipurpose 6061 Aluminum Rectangular Tube, 3/16" Wall Thickness, 2" High x 2" Wide
Catalog No. NC2899605 Supplier McMaster-Carr Supplier No. 6546K246FT
Multipurpose 6061 Aluminum Rectangular Tube, 3/16" Wall Thickness, 2" High x 2" Wide