Learn More
Perkin Elmer US LLC Aluminum Reference Material - Qty. 5
Catalog No. 502111483
Encompass_Preferred
PerkinElmer's Reference Materials ensure accurate thermogravimetric measurements for PerkinElmer's TA instruments. Magnetic Calibration Reference Standards are available as a kit as well as individually. PerkinElmer recommends the use of a fresh standard every time you calibrate your instruments, to avoid the effects of hysteresis and oxidation on the Curie Point of these standards.
Catalog No. 50-211-1483 Supplier Perkin Elmer US LLC Supplier No. N5380057
PerkinElmer's Reference Materials ensure accurate thermogravimetric measurements for PerkinElmer's TA instruments. Magnetic Calibration Reference Standards are available as a kit as well as individually. PerkinElmer recommends the use of a fresh standard every time you calibrate your instruments, to avoid the effects of hysteresis and oxidation on the Curie Point of these standards.