Learn More
Perkin Elmer US LLC Aluminum Std Expansion 10mm by 3 mm D
Catalog No. 502016360
Encompass_Preferred
The Aluminum Standatd Expansion 10 mm by 3 mm D is an aluminum reference material for the expansion calibration of the TMA 4000.
Catalog No. 50-201-6360 Supplier Perkin Elmer US LLC Supplier No. L8041600
The Aluminum Standatd Expansion 10 mm by 3 mm D is an aluminum reference material for the expansion calibration of the TMA 4000.