Learn More
TA Instruments Q50 Aluminum Sample Pans, 100ul, 12/PK
Supplier:  TA Instruments 952323.902
Encompass
100 ul Aluminum Sample Pans for th TGA Q500. These samples pans are also suitable for use with the TGA-HP and VTI-SA Sorption Analyzers. The pans are sold in packages of 12.
Catalog No. NC9924361