Learn More
TA Instruments Aluminum Sample Pans 80µL Pkg of 100, for Use in Discovery TGA 55, 550, 5500, Ambient to 600°C, Stainless Steel Bail Required P/N 957364.901
Catalog No. 5000580674 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Aluminum Sample Pans 80 µL Pkg Of 100 are for use in the Discovery TGA 55, 550, and 5500 over the temperature range of ambient to 600°C. A stainless steel bail P/N 957364.901 is required to use the aluminum sample pan in a TGA.
Catalog No. 50-005-80674 Supplier TA Instruments Supplier No. 957363.901
Waters TA Instruments Aluminum Sample Pans 80 µL Pkg Of 100 are for use in the Discovery TGA 55, 550, and 5500 over the temperature range of ambient to 600°C. A stainless steel bail P/N 957364.901 is required to use the aluminum sample pan in a TGA.