Learn More
Perkin Elmer US LLC Aluminum Sample Pans and Covers - Qty. 400
Catalog No. 502112120
Encompass_Preferred
The Aluminum Sample Pans and Covers are used with nonvolatile solid samples, such as polymers and pharmaceuticals. They are routinely used for applications such as polymer melting, crystallization, glass transitions of polymers, thermoplastics, and thermosets. Pans are crimped but not hermetically sealed.
Catalog No. 50-211-2120 Supplier Perkin Elmer US LLC Supplier No. 02190041
The Aluminum Sample Pans and Covers are used with nonvolatile solid samples, such as polymers and pharmaceuticals. They are routinely used for applications such as polymer melting, crystallization, glass transitions of polymers, thermoplastics, and thermosets. Pans are crimped but not hermetically sealed.