Learn More
TA Instruments Aluminum Sample Pans for SFI Samples, DSC Solid Fat Index Pans, Recessed Rim Prevents Wicking, Evaluates Edible Fats and Oils, Compatible with Standard DSC Lids, 200 Pack
Catalog No. 5000580665 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Aluminum Sample Pans For SFI Samples are DSC solid fat index pans designed specifically for evaluating edible fats and oils. The pans feature a recessed rim around the bottom to prevent wicking due to surface tension and are compatible with standard DSC lids.
Catalog No. 50-005-80665 Supplier TA Instruments Supplier No. 900870.901
Waters TA Instruments Aluminum Sample Pans For SFI Samples are DSC solid fat index pans designed specifically for evaluating edible fats and oils. The pans feature a recessed rim around the bottom to prevent wicking due to surface tension and are compatible with standard DSC lids.