Learn More
Glass Expansion Aluminum Sampler Cone Agilent
SDP
Catalog No. 502703005
Encompass_Preferred
Aluminum Sampler Cone. – For Agilent 7700/7800/7850/7900/8800/8900 ICP-MS. For lowest nickel background and laser ablation applications. Not for samples containing hydrofluoric acid (HF). Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For corrosive acids, including HF, use Pt Sampler with Ni Base, P/N 501907586. For high TDS samples causing rapid cone deposition, use Pt Sampler with Ni Base, P/N 501907585.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3005 Supplier Glass Expansion Supplier No. AT7701AL
Aluminum Sampler Cone. – For Agilent 7700/7800/7850/7900/8800/8900 ICP-MS. For lowest nickel background and laser ablation applications. Not for samples containing hydrofluoric acid (HF). Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For corrosive acids, including HF, use Pt Sampler with Ni Base, P/N 501907586. For high TDS samples causing rapid cone deposition, use Pt Sampler with Ni Base, P/N 501907585.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001