Learn More
Glass Expansion Aluminum Sampler Cone for Thermo Scientifc X Series/Q/RQ/TQ Series ICP-MS
SDP
Catalog No. NC1766768
Encompass_Preferred
Aluminum Sampler Cone, for Thermo Q/RQ/TQ and X Series ICP-MS. Ideal for lowest Ni background and laser ablation applications. Not for use with samples containing hydrofluoric acid (HF). Use a new Graphite Sampler Cone Gasket (TG5001) each time the cone is re-installed, even if it¿s the same cone. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For samples containing HF and other highly corrosive samples, see catalog number TG1048-Pt. For common applications with <5% acid matrix, non-HF, non-organic, see catalog number 501694614.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. NC1766768 Supplier Glass Expansion Supplier No. TG1021AL
Aluminum Sampler Cone, for Thermo Q/RQ/TQ and X Series ICP-MS. Ideal for lowest Ni background and laser ablation applications. Not for use with samples containing hydrofluoric acid (HF). Use a new Graphite Sampler Cone Gasket (TG5001) each time the cone is re-installed, even if it¿s the same cone. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For samples containing HF and other highly corrosive samples, see catalog number TG1048-Pt. For common applications with <5% acid matrix, non-HF, non-organic, see catalog number 501694614.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001