Learn More
DWK Life Sciences Kimble™ Aluminum Seal Manual Crimper
SDP
Catalog No. 12141437 Shop All DWK Life Sciences Products
Change view
Click to view available options
For Use With (Equipment):
Size 11 Aluminum Vial Seals
Size 13 Aluminum Vial Seals
Size 20 Aluminum Vial Seals
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
12-141-437 Size 11 Aluminum Vial Seals
12-141-438 Size 13 Aluminum Vial Seals
12-141-439 Size 20 Aluminum Vial Seals
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12-141-437 Supplier DWK Life Sciences Supplier No. 6990211
Add to Cart
Add to Cart
Manual hand crimper for aluminum seals and vials. Perfect for chromatography and general crimping needs.
  • Manual crimper for cap size 11 mm
  • Decrimper part number is 69904-11
TRUSTED_SUSTAINABILITY
Specifications
No. per Pack 1 Each
Type Manual Crimper
For Use With (Equipment) Size 11 Aluminum Vial Seals