Learn More
Strem, An Ascensus Company CAS# 7429-90-5. 100g. Aluminum shot (99.999%). MFCD00134029
SDP
Catalog No. 5090109521
Encompass
CAS# 7429-90-5. 100g. Aluminum shot (99.999%). MFCD00134029. Molecular Weight: 26.98. Molecular Formula: Al. Color/form: 6.35 x 6.35 mm cylinders Strem# 13-0044. http://www.strem.com/catalog/v/13-0044/
Catalog No. 50-901-09521 Supplier Strem, An Ascensus Company Supplier No. 130044100G
CAS# 7429-90-5. 100g. Aluminum shot (99.999%). MFCD00134029. Molecular Weight: 26.98. Molecular Formula: Al. Color/form: 6.35 x 6.35 mm cylinders Strem# 13-0044. http://www.strem.com/catalog/v/13-0044/