Learn More
Brady™ Aluminum Sign: REPORT ALL INJURIES OR UNSAFE CONDITIONS IMMEDIATELY
Catalog No. 19112540 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Aluminum
Catalog No. Material
19-112-540 Aluminum
1 options
Catalog No. 19-112-540 Supplier Brady™ Supplier No. 140819
  • $55.30
    $47.65 / Each of 1
    Save $7.65
    14% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Improve workplace safety with proper signage and markings. This Brady™ aluminum sign is a durable, clear and effective tool to raise awareness to workplace hazards.
  • Aluminum material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Aluminum signs provide the strength and durability of metal, but never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Specifications
Material Aluminum
Shape Rectangular
Languages English
Dimensions (L x W) 23.875 x 6 in.
Displayed Text REPORT ALL INJURIES OR UNSAFE CONDITIONS IMMEDIATELY
Product Type Safety Sign
Color Blue on White
WARNING: Cancer - www.P65Warnings.ca.gov
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.