Learn More
Brady™ Aluminum Sign: EVERY PERSON HAS THE RESPONSIBILITY AND AUTHORITY TO STOP WORK IN AN UNSAFE SITUATION
Shop All Brady Corporation ProductsDescription
- Aluminum material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- Aluminum signs provide the strength and durability of metal, but never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Specifications
Specifications
| Material | Aluminum |
| Shape | Rectangular |
| Languages | English |
| Dimensions (L x W) | 23.875 x 6 in. |
| Displayed Text | EVERY PERSON HAS THE RESPONSIBILITY AND AUTHORITY TO STOP WORK IN AN UNSAFE SITUATION |
| Product Type | Safety Sign |
| Color | Blue on White |
Safety and Handling
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.