Learn More
Brady™ Aluminum Sign: NON-POTABLE WATER
Catalog No. 19106773 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
12 x 9 in.
Catalog No. Dimensions (L x W)
19-106-773 12 x 9 in.
1 options
Catalog No. 19-106-773 Supplier Brady™ Supplier No. 17582
Improve workplace safety with proper signage and markings. This Brady™ aluminum sign is a durable, clear and effective tool to raise awareness to workplace hazards.
  • Aluminum material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Aluminum signs provide the strength and durability of metal, but never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Specifications
Material Aluminum
Shape Rectangular
Dimensions (L x W) 12 x 9 in.
Displayed Text NON-POTABLE WATER
Color Red on White