Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Aluminum Utility Panels, Number and Letter: 12 x 1.75 in

Catalog No. 19112816 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
12 x 1.75 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-112-816 12 x 1.75 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-112-816 Supplier Brady™ Supplier No. 97199
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Improve workplace safety with proper signage and markings. This Brady™ aluminum sign panel is a durable, clear and effective tool to raise awareness to workplace hazards.

  • Material: Aluminum (0.020 in. thick)
  • Design: Rugged yet flexible to conform to pole curves
  • Mounting: Post mounted
  • Loading Orientation: Horizontal or vertical
  • Content: Blank

Specifications

Material Aluminum
Mounting Type Post Mount
Shape Rectangular
Languages English
Dimensions (L x W) 12 x 1.75 in.
Displayed Text LOOK OUT FOR LIFT TRUCKS. ACCIDENT WILL CAUSE SEVERE INJURY OR DEATH.
Product Type Safety Sign
Color Silver

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.