Learn More
Description
Specifications
Specifications
| CAS | 1344-00-9 |
| Quantity | 250 g |
| MDL Number | MFCD00131613 |
| Synonym | Aluminum sodium silicate; P 820 A |
| Percent Purity | 8%Na2O basis (based on calcined substance); 82%SiO2 basis (based on calcined substance); 9.5%Al2O3 basis (based on calcined substance) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.