Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum silicate

Catalog No. AA14231A3
Click to view available options
Quantity:
500 g
2 kg

CAS: 12428-46-5 | Al2H4O9Si2 | 258.16 g/mol

Aluminum silicate is used to prepare aluminum-silicate polymer composite (PASiC), which is an inorganic coagulant used in water treatment. It is used as a catalyst for the pyrolysis of rice husk to crude bio -oil, which upgraded to supercritical ethanol. Further, it is used as filler in paper, rubber and in paints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12428-46-5
Molecular Formula Al2H4O9Si2
Molecular Weight (g/mol) 258.16
MDL Number MFCD00062311
InChI Key NLYAJNPCOHFWQQ-UHFFFAOYSA-N
Synonym kaolin, china clay, argilla, glomax, porcelain clay, bolus alba, white bole, hydrated aluminum silicate, kaolin clay slurry, alphagloss
PubChem CID 56841936
IUPAC Name oxo-oxoalumanyloxy-[oxo(oxoalumanyloxy)silyl]oxysilane;dihydrate
SMILES O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O

Specifications

Physical Form Powder
Quantity 2 kg
Solubility Information Insoluble in water.
Formula Weight 258.16 (222.13anhydrous)
Chemical Name or Material Aluminum silicate
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.