Learn More
Glass Expansion Aluminum Skimmer Cone PerkinElmer NexION 300/350/1000/1100/2000
SDP
Catalog No. 502702954
Encompass_Preferred
Aluminum Skimmer Cone ¿ Aluminum skimmer cone for PerkinElmer NexION 1000/2000/300/350 ICP-MS. For lowest nickel background and laser ablation applications. Not recommended for samples containing HF. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection. Use skimmer ConeGuard to protect cone threads during cleaning, 70-803-1026.For standard samples and routine operation, we recommend Nickel Skimmer Cones, 501907685. For corrosive samples, including HF, we recommend Platinum Skimmer Cones, see catalog number 501907688.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2954 Supplier Glass Expansion Supplier No. PE3012AL
Aluminum Skimmer Cone ¿ Aluminum skimmer cone for PerkinElmer NexION 1000/2000/300/350 ICP-MS. For lowest nickel background and laser ablation applications. Not recommended for samples containing HF. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection. Use skimmer ConeGuard to protect cone threads during cleaning, 70-803-1026.For standard samples and routine operation, we recommend Nickel Skimmer Cones, 501907685. For corrosive samples, including HF, we recommend Platinum Skimmer Cones, see catalog number 501907688.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001