Learn More
Glass Expansion Aluminum Skimmer Cone x-lens
SDP
Catalog No. 502703001
Encompass_Preferred
Aluminum Skimmer Cone for x-lens – For Agilent 7900/8900 ICP-MS. For lowest nickel background and laser ablation applications. Not for samples containing hydrofluoric acid (HF). Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. Use Skimmer ConeGuard (70-803-1004) to protect cone threads during cleaning. When not in use, keep cones stored in original packaging for protection.For corrosive acids, including HF, use Pt Skimmer with Cu Base for x-lens, P/N 501907594, must be mounted on brass adaptor skimmer base. For organic analysis, use Pt Skimmer with Ni Base for x-lens, P/N 501907595, which must be mounted on brass adaptor skimmer base.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3001 Supplier Glass Expansion Supplier No. AT7902XAL
Aluminum Skimmer Cone for x-lens – For Agilent 7900/8900 ICP-MS. For lowest nickel background and laser ablation applications. Not for samples containing hydrofluoric acid (HF). Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. Use Skimmer ConeGuard (70-803-1004) to protect cone threads during cleaning. When not in use, keep cones stored in original packaging for protection.For corrosive acids, including HF, use Pt Skimmer with Cu Base for x-lens, P/N 501907594, must be mounted on brass adaptor skimmer base. For organic analysis, use Pt Skimmer with Ni Base for x-lens, P/N 501907595, which must be mounted on brass adaptor skimmer base.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001