Learn More
Electron Microscopy Sciences Aluminum 10 Slide Tray
SDP
Catalog No. 50931149
Encompass_Preferred
Aluminum slide trays have elevated ridges to separate 76 x 26mm slides and finger holes for easy removal, ideal for drying fresh mounts.
Capacity: 10 slides
Dimensions: 13'L x 4"W
Catalog No. 50-931-149 Supplier Electron Microscopy Sciences Supplier No. 7150610
Aluminum slide trays have elevated ridges to separate 76 x 26mm slides and finger holes for easy removal, ideal for drying fresh mounts.
Capacity: 10 slides
Dimensions: 13'L x 4"W