Learn More
Electron Microscopy Sciences Aluminum 20 Slide Tray
SDP
Catalog No. 50931150
Encompass_Preferred
Aluminum slide trays have elevated ridges to separate 76 x 26mm slides and finger holes for easy removal, ideal for drying fresh mounts.
Capacity: 20 slides
Dimensions: 13"L x 7-1/2"W
Catalog No. 50-931-150 Supplier Electron Microscopy Sciences Supplier No. 7150620
Aluminum slide trays have elevated ridges to separate 76 x 26mm slides and finger holes for easy removal, ideal for drying fresh mounts.
Capacity: 20 slides
Dimensions: 13"L x 7-1/2"W