Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Aluminum Sodium Sulfate, Dodecahydrate
SCI_ED

Catalog No. S25136 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Lab Grade - Crystals
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25136 500 g Lab Grade - Crystals
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S25136
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 10102-71-3
Molecular Formula AlNaO8S2
Molecular Weight (g/mol) 242.083
InChI Key GJPYYNMJTJNYTO-UHFFFAOYSA-J
Synonym sodium aluminum sulfate, aluminum sodium sulfate, aluminum sodium disulfate, unii-0cm6a697vv, sodium aluminum sulfate, aluminum sodium bis sulphate, hsdb 571, sulfuric acid, aluminum sodium salt 2:1:1, aluminum sodium sulfate, naal so4 2, aluminum sodium sulfate alna so4 2
PubChem CID 24939
IUPAC Name aluminum;sodium;disulfate
SMILES [O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Na+].[Al+3]

Specifications

Quantity 500 g
Linear Formula AlNa(SO4)2
Formula Weight 458.29
Grade Laboratory
Packaging Lab Grade - Crystals
Density 1.67g/mL
Chemical Name or Material Aluminum Sodium Sulfate, Dodecahydrate
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.