Learn More
Medchemexpress LLC Aluminum stearate (octadecanoate aluminum) | 637-12-7 | MFCD00036389 | 293.47 g/mol | C18H36O2·1/3Al | 500 G
SDP
Catalog No. 5000283126
Encompass_Preferred
Aluminum stearate (octadecanoate aluminum) is a fatty-acid metal salt used as a thickener and lubricant in cosmetic and industrial formulations. Supplied as a white to off-white solid, it is typically used to increase viscosity and improve texture in oil-based products.
Catalog No. 50-002-83126 Supplier Medchemexpress LLC Supplier No. HYW133900500G
Aluminum stearate (octadecanoate aluminum) is a fatty-acid metal salt used as a thickener and lubricant in cosmetic and industrial formulations. Supplied as a white to off-white solid, it is typically used to increase viscosity and improve texture in oil-based products.