Learn More
Aluminum sulfate, anhydrous, 99.99% (metals basis)
Description
Aluminum sulfate acts as a flocculating agent in the purification of drinking water, and in waste water treatment plants. It acts as a mordant in dyeing, printing, textiles and also used in paper manufacturing. It is a waterproofing agent and accelerator in concrete. Further, it is used as a foaming agent in fire fighting foam, photographic film and in photochemicals. It is also used in styptic pencils, pain relief and in dentistry for gingival retraction cords.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Specifications
Specifications
| Melting Point | 770°C |
| pH | 3.5 |
| Physical Form | Powder |
| Quantity | 25 g |
| Merck Index | 14,366 |
| Solubility Information | Soluble in water. Slightly soluble in alcohol and dilute mineral acids. Insoluble in ethanol. |
| Formula Weight | 342.15 |
| Percent Purity | 99.99% |
| Sensitivity | Hygroscopic |
| Density | 2.71 g/mL |
| Show More |
Safety and Handling
| Hazard Category | H318 |
| Hazard Statement | GHS H Statement H318-H335-H315 Causes serious eye damage. May cause respiratory irritation. Causes skin irritation. |
| Precautionary Statement | P280-P305+P351+P338-P310 |
| EINECSNumber | 233-135-0 |
| RTECSNumber | BD1700000 |
| TSCA | Yes |
| storageNote1 |
|
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.