Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Aluminum Sulfate, Hydrate
SCI_ED

Catalog No. S25137 Shop All Fisherbrand Products
Change view
Click to view available options
500g; Lab Grade
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S25137 500g; Lab Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25137

missing translation for 'validMainframeMsg'

Paper sizing

This product is an Educational Grade Chemical — no C of A is available.

Specifications

CAS 7784-31-8
Quantity 500 g
Molecular Formula Al2H36O30S3
Linear Formula Al2(SO4)3·18H2O
Synonym aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18
InChI Key AMVQGJHFDJVOOB-UHFFFAOYSA-H
SMILES O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]
IUPAC Name dialuminum;trisulfate;octadecahydrate
Molecular Weight (g/mol) 666.401
PubChem CID 22377415
ChEBI CHEBI:74779
Formula Weight 666.42
Grade Laboratory
Packaging Lab Grade
Chemical Name or Material Aluminum Sulfate, Hydrate
Show More Show Less
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.