Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Sulfate, Purified, Reagents

Catalog No. 01337507
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-507 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-507 Supplier Reagents Holdings Llc Supplier No. C2128500500A5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aluminum Sulfate, Purified, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 17927-65-0
Molecular Formula Al2O12S3
Molecular Weight (g/mol) 342.13
MDL Number MFCD00149136
InChI Key DIZPMCHEQGEION-UHFFFAOYSA-H
Synonym Dialuminum trisulfate hydrate
IUPAC Name dialuminium(3+) trisulfate
SMILES [Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color White to Off-White
Physical Form Solid
Quantity 500 g
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.6
Chemical Name or Material Aluminum Sulfate, Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.