Learn More
Supertek™ Aluminum Superior Grade 2048 Hz 'C' Tuning Fork
GREENER_CHOICESCI_ED
Catalog No. S81850 Shop All Supertek Scientific Products
Change view
Click to view available options
Frequency:
2048 Hz
Catalog No. Frequency
S81850 2048 Hz
1 options
This item is not returnable. View return policy
Catalog No. S81850 Supplier Supertek™ Supplier No. PH36121/12
Add to Cart
Add to Cart
This item is not returnable. View return policy
This Supertek™ Aluminum Superior Grade 2048 Hz 'C' Tuning Fork, three octaves above middle 'C' has been individually tested for accuracy.
  • Tuning fork is stamped with frequency and tone
Specifications
Product Type Tuning Fork
Material Aluminum
Frequency 2048 Hz