Learn More
Supertek™ Aluminum Superior Grade 512 Hz 'C' Tuning Fork
GREENER_CHOICESCI_ED
Catalog No. S81848 Shop All Supertek Scientific Products
Change view
Click to view available options
Frequency:
512 Hz
Catalog No. Frequency
S81848 512 Hz
1 options
This item is not returnable. View return policy
Catalog No. S81848 Supplier Supertek™ Supplier No. PH36121/10
Add to Cart
Add to Cart
This item is not returnable. View return policy
This Supertek™ Aluminum Superior Grade 512 Hz 'C' Tuning Fork, one octave above middle 'C,' has been individually tested for accuracy.
  • Tuning fork is stamped with frequency and tone
Specifications
Product Type Tuning Fork
Material Aluminum
Frequency 512 Hz