Learn More
Electron Microscopy Sciences Scotch™ Aluminum Tape, 1" wide 18 YD
SDP
Catalog No. 5028524
Encompass_Preferred
The conductive tape is used in scanning electron microscopy specimen mounting. Aluminum foil tape (Scotch 1170) with conductive adhesive. 3" core and foil thickness: 2 mils. Total thickness: 4mils (.102mm), adhesion: 35oz/in, electrical resistance through adhesive: .01ohm/in(.07ohm/cm)
Width: 1"
Catalog No. 50-285-24 Supplier Electron Microscopy Sciences Supplier No. 77798
The conductive tape is used in scanning electron microscopy specimen mounting. Aluminum foil tape (Scotch 1170) with conductive adhesive. 3" core and foil thickness: 2 mils. Total thickness: 4mils (.102mm), adhesion: 35oz/in, electrical resistance through adhesive: .01ohm/in(.07ohm/cm)
Width: 1"