Learn More
Electron Microscopy Sciences Aluminum Tissue Mounting Stub
SDP
Catalog No. 5029359
Encompass_Preferred
Machined from aluminum rods with one end tapered to 4.8 mm in diameter. The specimen can be oriented and mounted to the tip with glue and it is ready for sectioning. The body size is "00" and it measures 11 mm long.
Catalog No. 50-293-59 Supplier Electron Microscopy Sciences Supplier No. 7014505
Machined from aluminum rods with one end tapered to 4.8 mm in diameter. The specimen can be oriented and mounted to the tip with glue and it is ready for sectioning. The body size is "00" and it measures 11 mm long.