Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum titanium oxide, 99.5% (metals basis)

Catalog No. AA1448414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1448414 25 g
AA1448422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1448414 Supplier Thermo Scientific Chemicals Supplier No. 01448414
Add to Cart

CAS: 12004-39-6 | Al2O5Ti | 181.825 g/mol

Aluminum titanium oxide is used in refractory industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12004-39-6
Molecular Formula Al2O5Ti
Molecular Weight (g/mol) 181.825
MDL Number MFCD00064749
InChI Key CNRZQDQNVUKEJG-UHFFFAOYSA-N
Synonym aluminum titanate, powder, oxo-bis oxoalumanyloxy titanium, trioxo-2,4-dioxa-1,5-dialumina-3-titanapentane, aluminum titanate, nanopowder, <25 nm particle size bet trace metals basis
PubChem CID 16213786
IUPAC Name oxo-bis(oxoalumanyloxy)titanium
SMILES O=[Al]O[Ti](=O)O[Al]=O

Specifications

Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 181.86
Percent Purity 99.5%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Aluminum titanium oxide
Hazard Category Warning
Hazard Statement GHS H Statement
H335
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P304+P340-P312-P405-P403+P233-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Store in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 234-456-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.