Learn More
Labrepco Inc ALUMINUM VERTICAL RACKS FOR CH
SDP
Supplier:  Labrepco Inc CFA132
Encompass_Preferred
Rack, Vertical; Labrepco; For chest freezer and liquid nitrogen tanks; Aluminum; For standard 2 in. boxes; Dimension (D x W x H): 5.5 x 5.625 x 28.3125 in.or 140 x 143 x 719mm; Box capacity: 13
Catalog No. 50-902-0149