Learn More
Labrepco Inc ALUMINUM VERTICAL RACKS FOR CH
SDP
Catalog No. 509020147
Encompass_Preferred
Rack, Vertical; Labrepco; For chest freezer and liquid nitrogen tanks; Aluminum; For standard 2 in. boxes; Dimension (D x W x H): 5.5 x 5.625 x 24.187 in.or 140 x 143 x 615mm; Box capacity: 11
Catalog No. 50-902-0147 Supplier Labrepco Inc Supplier No. CFA112
Rack, Vertical; Labrepco; For chest freezer and liquid nitrogen tanks; Aluminum; For standard 2 in. boxes; Dimension (D x W x H): 5.5 x 5.625 x 24.187 in.or 140 x 143 x 615mm; Box capacity: 11