Learn More
Khk Enterprises Inc ALUMINUM WEIGHING DISH 12X3X4M
Catalog No. NC9935594
Encompass
A pure aluminum sample vessel and weighing dish that is ideal as a Sample carrier for weighing equipment or as a container in MicroBalance applications where light weight and material purity are a must. Product dimension are 13x3x4mm (.0005 thick)
Catalog No. NC9935594 Supplier Khk Enterprises Inc Supplier No. C0290410/3
A pure aluminum sample vessel and weighing dish that is ideal as a Sample carrier for weighing equipment or as a container in MicroBalance applications where light weight and material purity are a must. Product dimension are 13x3x4mm (.0005 thick)