Learn More
Khk Enterprises Inc ALUMINUM WEIGHING DISHES 100BX
Catalog No. NC9032543
Encompass
A pure aluminum sample vessel and weighing dish that is ideal as a Sample carrier for weighing equipment or as a container in MicroBalance applications where light weight and material purity are a must. Product dimension are 13x3x4mm (.0005 thick)
Catalog No. NC9032543 Supplier Khk Enterprises Inc Supplier No. C0290410
A pure aluminum sample vessel and weighing dish that is ideal as a Sample carrier for weighing equipment or as a container in MicroBalance applications where light weight and material purity are a must. Product dimension are 13x3x4mm (.0005 thick)