Learn More
Khk Enterprises Inc Aluminum Weighing Pans, 100 per Pack
Catalog No. NC9630673
Encompass
Pan, Sample, Aluminum, for Microbalances, 0.040mm Thick, 22L x 7W x 7mmH, 70 MG
Catalog No. NC9630673 Supplier Khk Enterprises Inc Supplier No. C0290412
Pan, Sample, Aluminum, for Microbalances, 0.040mm Thick, 22L x 7W x 7mmH, 70 MG